Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ICMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585576 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585576 Supplier Invitrogen™ Supplier No. PA585576
  • $582.00
    $472.65 / Each of 1

    Save $109.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Pig (100%). Positive Control: HeLa membrane extract (unboiled). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the third of three enzymes that post translationally modify isoprenylated C-terminal cysteine residues in certain proteins and target those proteins to the cell membrane. This enzyme localizes to the endoplasmic reticulum. Alternative splicing may result in other transcript variants, but the biological validity of those transcripts has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICMT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Icmt
Gene Accession No. O60725, Q9EQK7, Q9WVM4
Gene Alias 1700008E11Rik; C80758; farnesyl cysteine carboxyl methyltransferase; fcmt; Gm13095; HSTE14; Icmt; isoprenylcysteine carboxyl methyltransferase; Isoprenylcysteine carboxylmethyltransferase; MGC39955; MST098; MSTP098; OTTMUSG00000010406; pcCMT; PCMT; PPMT; prenylated protein carboxyl methyltransferase; prenylcysteine carboxyl methlytransferase; prenylcysteine carboxyl methyltransferase; protein-S isoprenylcysteine O-methyltransferase; protein-S-isoprenylcysteine O-methyltransferase; STE14
Gene Symbols Icmt
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ICMT. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170818, 23463, 57295
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.