Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS (CD278) Armenian Hamster anti-Human, Mouse, Alexa Fluor™ 488, Clone: C398.4A, BD Phrmingen™

Catalog No. BDB569525 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569525 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569525 Supplier BD Biosciences Supplier No. 569525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Armenian Hamster Monoclonal Antibody

ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human∣Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICOS (CD278)
Applications Flow Cytometry
Classification Monoclonal
Clone C398.4A
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115
Host Species Armenian Hamster
Immunogen Mouse D10.G4.1 T-cell clone
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29851
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.