Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICP Standard, 100 ppm Silver in dilute HNO3, Ricca Chemical
SDP

Catalog No. RPAG100N100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RPAG100N100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RPAG100N100 Supplier Ricca Chemical Company Supplier No. RPAG100N100B
Add to Cart
Add to Cart

Single Element ICP Standard

Chemical Identifiers

CAS 7440-22-4 , 7697-37-2 , 7732-18-5
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Packaging Amber Poly Bottle
CAS Min % 96.9, 3.09, 0.0098
CAS Max % 96.9, 3.09, 0.0098
Density 1.02
Physical Form Liquid
Color Colorless
Melting Point 0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material ICP Standard, 100 ppm Silver in Dilute HNO3
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.