Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho ID-Micro Typing System™ Reagents
SDP

Catalog No. OKMTS9610
Change view
Click to view available options
Quantity:
0.5 mL
1 mL
100 mL
Type:
ID-Micro Typing System Diluent 2
ID-Micro Typing System Diluent 2 Plus
ID-Micro Typing System Reagent Dispenser
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
OKMTS9610 0.5 mL ID-Micro Typing System Reagent Dispenser
OKMTS9612 1 mL ID-Micro Typing System Reagent Dispenser
OKMTS9230 100 mL ID-Micro Typing System Diluent 2
OKMTS9330 100 mL ID-Micro Typing System Diluent 2 Plus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. OKMTS9610 Supplier QuidelOrtho Supplier No. MTS9610
Add to Cart
Add to Cart

Reagents for the QuidelOrtho Clinical Diagnostics ID-Micro Typing System Gel Workstation

  • Suitable for a broad range of blood bank applications
  • Use proven serologic methods

Antibody screening and identification, ABO blood grouping and Rh phenotyping, Compatibility testing, Reverse serum grouping and antigen typing

Specifications

Type ID-Micro Typing System Reagent Dispenser
For Use With (Application) For ID-Micro typing system™ gel workstation, Antibody screening and identification, ABO Blood grouping and Rh phenotyping, compatibility testing and reverse Serum grouping and Antigen typing
Quantity 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.