Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID3 Mouse anti-Human, Clone: OTI8B3, lyophilized, TrueMAB™
SDP

Catalog No. 501659771
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-165-9771 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-165-9771 Supplier OriGene Supplier No. CF500713
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Members of the Id family of basic helix-loop-helix (bHLH) proteins include ID1, ID2, ID3 and ID4. They are ubiquitously expressed and dimerize with members of the class A and B HLH proteins. Due to the absence of the basic region, the resulting heterodimers cannot bind DNA. The ID-type proteins thus appear to negatively regulate DNA binding of bHLH proteins. Since ID1 inhibits DNA binding of E12 and Myo D, it apparently functions to inhibit muscle-specific gene expression. Under conditions that facilitate muscle cell differentiation, the ID protein levels fall, allowing E12 and/or E47 to form heterodimers with Myo D and myogenin, which in turn activate myogenic differentiation. It has been shown that expression of each of the ID proteins is strongly dependent on growth factor activation and that reduction of ID mRNA levels by antisense oligonucleotides leads to a delayed reentry of arrested cells into the cell cycle following growth factor stimulation.

Specifications

Antigen ID3
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI8B3
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ID3
Gene Accession No. Q02535
Gene Alias HEIR-1, bHLHb25
Gene Symbols ID3
Host Species Mouse
Immunogen Full length human recombit protein of human ID3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3399
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.