Learn More
TARGETMOL CHEMICALS INC IDE1 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3162
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IDE1 can induce definitive endoderm from embryonic stem cells. It has been shown to induce the differentiation of Sox17+/FoxA2+-expressing pancreatic progenitors from human and mouse embryonic stems cells (EC50 125.5 nM in vitro) by activating the TGF-(beta) signaling pathway. IDE1-derived endodermal cells injected into E8.75 mouse embryos ex vivo have been shown to incorporate into the developing gut tube contributing to its formation. purity: 98%
Catalog No. 50-258-0358
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.