Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Idebenone, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18611007 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-007 1 kg Amber Glass Bottle
18-611-010 5 g Amber Glass Bottle
18-611-009 25 g Amber Glass Bottle
18-611-008 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-007 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I20161KGBL
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 58186-27-9
Molecular Formula C19H30O5
Molecular Weight (g/mol) 338.44
InChI Key JGPMMRGNQUBGND-UHFFFAOYSA-N
IUPAC Name 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
SMILES COC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O

Specifications

CAS Max % 1
Melting Point 52.0° to 55.0°C
Loss on Drying 0.02
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 0.985
Grade Reagent

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.