Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega IdeS Protease

Catalog No. PRV7511 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
5 x 5000 U
5000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV7511 5000 U
PRV7515 5 x 5000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV7511 Supplier Promega Supplier No. V7511
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IdeS Protease is an immunoglobulin-degrading enzyme from Streptococcus pyogenes (IdeS). IdeZ Protease is an immunoglobulin-degrading enzyme from Streptococcus equi subspecies zooepidemicus.

  • Antibody Fragmentation and Characterization, Simplified
  • Highly specific IgG cleavage
  • Rapid generation of highly pure fragments
  • Frozen protease is supplied at a concentration of 50u/μL

Specifications

Content And Storage -30°C to -10°C
Quantity 5000 U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.