Learn More
Leap PAL Parts Idex valve with actuator, 7 port, 6 position, 1/16"", 0.20mm, Max pressure: 15,000 psi
Supplier:  Leap PAL Parts ID.MXT715105
Encompass
Valve with actuator, Idex/Rheodyne, 7 port, 6 position, 1/16", 0.20mm, Max pressure: 15,000 psi
Catalog No. 50-187-1403
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.