Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDH1 Monoclonal Antibody (A11-B4)
GREENER_CHOICE

Catalog No. PIMA541084
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541084 Supplier Invitrogen™ Supplier No. MA541084
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IDH1 Monoclonal Antibody for IHC (P), Western Blot

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP(+)-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP(+)-dependent isocitrate dehydrogenase found in the cytoplasm and peroxisomes. It contains the PTS-1 peroxisomal targeting signal sequence. The presence of this enzyme in peroxisomes suggests roles in the regeneration of NADPH for intraperoxisomal reductions, such as the conversion of 2, 4-dienoyl-CoAs to 3-enoyl-CoAs, as well as in peroxisomal reactions that consume 2-oxoglutarate, namely the alpha-hydroxylation of phytanic acid. The cytoplasmic enzyme serves a significant role in cytoplasmic NADPH production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A11-B4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene IDH1
Gene Accession No. O75874
Gene Alias AI314845; AI788952; cb876; corneal epithelial crystallin; cytosolic NADP+-dependent isocitrate dehydrogenase; cytosolic NADP-isocitrate dehydrogenase; cytosolic NADP-isocitrate dehydrongenase; E030024J03Rik; epididymis luminal protein 216; epididymis secretory protein Li 26; fm90e09; HEL-216; HEL-S-26; ICDH; Id-1; IDCD; IDH; Idh1; Idh-1; IDP; Idpc; im:7143416; isocitrate dehydrogenase (NAD(+)) IDH1; isocitrate dehydrogenase (NADP(+)) 1; isocitrate dehydrogenase (NADP(+)) 1, cytosolic; Isocitrate dehydrogenase [NAD] subunit 1, mitochondrial; Isocitrate dehydrogenase [NADP] cytoplasmic; isocitrate dehydrogenase 1 (NADP+); isocitrate dehydrogenase 1 (NADP+), soluble; Isocitrate dehydrogenase 1, soluble; Isocitric dehydrogenase; N2690; NAD(+)-specific ICDH; NADP(+)-specific ICDH; NADP+-specific ICDH; NADP-cICDH; NADP-dependent isocitrate dehydrogenase, cytosolic; NADP-dependent isocitrate dehydrogenase, peroxisomal; OTTHUMP00000206464; OTTHUMP00000206465; OTTHUMP00000206466; oxalosuccinate decarboxylase; PICD; wu:fm90e09; YNL037C
Gene Symbols IDH1
Host Species Mouse
Immunogen Synthetic peptide within Human IDH1 aa 101-150.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100006589, 3417
Target Species Human, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.