Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IDH2 Monoclonal Antibody (GT673)
GREENER_CHOICE

Catalog No. PIMA517271 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517271 Supplier Invitrogen™ Supplier No. MA517271
  • $635.00
    $498.65 / Each of 1

    Save $136.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Recommended positive controls: HepG2, Jurkat, Raji, K562, THP-1, NCI-H929, mouse brain, rat brain, DDDDK-tagged IDH2-transfected 293T. Predicted reactivity: Mouse (97%), Rat (96%), Zebrafish (89%), Xenopus laevis (91%), Pig (96%), Chicken (95%), Rhesus Monkey (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NADas the electron acceptor and the other NADP. Five isocitrate dehydrogenases have been reported: three NAD-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP-dependent isocitrate dehydrogenase found in the mitochondria. It plays a role in intermediary metabolism and energy production. This protein may tightly associate or interact with the pyruvate dehydrogenase complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDH2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT673
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Idh2
Gene Accession No. P48735, P54071, P56574
Gene Alias D2HGA2; E430004F23; ICD-M; IDH; Idh2; Idh-2; IDHM; IDP; IDPM; isocitrate dehydrogenase (NADP(+)) 2; isocitrate dehydrogenase (NADP(+)) 2, mitochondrial; isocitrate dehydrogenase [NADP], mitochondrial; isocitrate dehydrogenase 2 (NADP+), mitochondrial; mNADP-IDH; NADP(+)-specific ICDH; NADP+-specific ICDH; NADP+-specific isocitrate dehydrogenase; NADPH-specific isocitrate dehydrogenase; oxalosuccinate decarboxylase
Gene Symbols Idh2
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human IDH2. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269951, 3418, 361596
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.