Learn More
Epigentek Inc IDO2 Polyclonal Antibody (50 ul)
SDP
Supplier:  Epigentek Inc A56410050
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Indoleamine 2,3-dioxygenase 2 protein (1-407AA). Tested Applications: ELISA, IHC, IF. Reactivity: Human. Alternative Names: Indoleamine 2, 3-dioxygenase-like protein 1, Indoleamine-pyrrole 2, 3-dioxygenase-like protein 1, INDOL1, IDO2.
Catalog No. 50-122-6931
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.