Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biocartis Idylla™ Console

Catalog No. 23555109
Change view
Click to view available options
Fully Automated Molecular Diagnostics System Console
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-555-109 Fully Automated Molecular Diagnostics System Console
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-555-109 Supplier Biocartis Supplier No. P1010

Fully automated, PCR-based Biocartis™ Idylla™ system offers fast and easy access to molecular Biomarker information. Compact console features touch-screen interface and enables laboratories to perform a broad range of applications.

Features:

  • Works on-demand in virtually any laboratory setting, allowing even de-centralized laboratores to rapidly report results
  • Covers the entire process from sample to result in 90 to 150 minutes
  • Allows laboratories to report same-day results, so physicians have the information needed to make timely decisions
  • Ease of use: Requires less than 2 minutes of operator time to run a sample
  • Fully integrated: Eliminates the need to manual sample preparation, addition of buffers or master-mix, pre-mixing of reagents or centrifugation
  • Multiplexing capability: Can detect up to 30 molecular targets in standard mode
  • Cartridge seal prevents spread of any DNA amplification products that might cause contamination of the laboratory equipment or environment
  • Built-in barcode scanner
  • LIS and remote connectivity capability

Physical dimensions:

  • w: 254mm / 10in
  • h: 506mm  /19.92in
  • d: 254mm / 10in
  • weight: 5.2kg

Operating conditions:

  • Temperature: 15 to 30°C
  • Humidity: 40 to 60% RH
  • Pressure: 80 to 106kPa

Power requirements:

  • Power supply: 100 to 200 V; 50/60Hz
  • Average power consumption: 100W
  • Noise level: Max. 49dB(A)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.