Learn More
Invivogen iE-DAP, 5 mg
SDP
Supplier:  Invivogen TLRLDAP
Encompass
gamma-D-Glu-mDAP (iE-DAP) is a dipeptide present in the peptidoglycan (PGN) of a subset of bacteria that include Gram-negative bacilli and particular Gram-positive bacteria such as Bacillus subtilis and Listeria monocytogenes.
Catalog No. NC9716316
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.