Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich IEICO ≥ 99 %

Catalog No. 913006100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-300-6100MG 100 mg
91-300-6250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 91-300-6100MG Supplier Sigma Aldrich Supplier No. 913006100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

IEICO is a high-performance narrow-bandgap non-fullerene acceptor (NFA) with a complementary absorption spectrum to many of the best medium to high bandgap OPV devices. This feature was used to create high-efficiency multi-layer tandem solar cells, reaching a PCE of nearly 14%. Over 11% PCE can be reached in single junction cells.

Specifications

CAS 2055812-53-6
Molecular Formula C114H118N4O4S4
Linear Formula C114H118N4O4S4
Quantity 100 mg
Synonym 2,2 -((2 Z,2 Z)-((5,5 -bis(4,4,9,9-tetrakis(4-hexylphenyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b]dithiophene-2,7-diyl)bis(4-((2- ethylhexyl)oxy)thiophene-5,2-diyl))bis(methanylylidene))bis-(3-oxo-2,3-dihydro-1 H-indene-2,1-diylidene))dimalononitrile; 2,2 -[[4,4,9,9-tetrakis(4-hexylphenyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b]dithiophene-2,7-diyl]bis[[4-[(2-ethylhexyl)oxy]-5,2-thiophenediyl]-(Z)-methylidyne(3-oxo-1 H-indene-2,1(3 H)-diylidene)]]bis-propanedinitrile
Molecular Weight (g/mol) 1736.44
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.