Learn More
TARGETMOL CHEMICALS INC IEM 1754 2HBR 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6134
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IEM 1754 2HBr (IEM 1754 dihydrobromide) is a selective AMPA/kainate receptor blockers for GluR1 and GluR3 with IC50 of 6 uM. purity: 99%
Catalog No. 50-257-9529
Only null left
Edge
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.