Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFITM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590244
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590244 Supplier Invitrogen™ Supplier No. PA590244
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNHIVQTFSP VNSGQPPNYE MLKEEQEVAM LGVPHNPAPP MSTVIHIRSE TSVPDHVVWS LFNTLFMNTC CLGFIAFAYS VKSRDRKMVG DVTGAQAYAS; Positive Samples: HeLa; Cellular Location: Cell membrane, Single-pass membrane protein.

IFN-induced antiviral protein which inhibits the entry of viruses to the host cell cytoplasm, permitting endocytosis, but preventing subsequent viral fusion and release of viral contents into the cytosol. Active against multiple viruses, including influenza A virus, SARS coronavirus (SARS-CoV), Marburg virus (MARV), Ebola virus (EBOV), Dengue virus (DNV), West Nile virus (WNV), human immunodeficiency virus type 1 (HIV-1) and vesicular stomatitis virus (VSV). Can inhibit: influenza virus hemagglutinin protein-mediated viral entry, MARV and EBOV GP1,2-mediated viral entry, SARS-CoV S protein-mediated viral entry and VSV G protein-mediated viral entry. Induces cell cycle arrest and mediates apoptosis by caspase activation and in p53-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFITM2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene IFITM2
Gene Accession No. Q01629
Gene Alias 1-8D; dispanin subfamily A member 2c; DSPA2c; IFITM2; ifm2; interferon induced transmembrane protein 2; interferon-induced transmembrane protein 2; Interferon-inducible protein 1-8D
Gene Symbols IFITM2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human IFITM2 (NP_0064262).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10581
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.