Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFITM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558715
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558715 Supplier Invitrogen™ Supplier No. PA558715
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SIKARDQKVV GDLEAARRFG SKAKC Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.

Plays a role in bone mineralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFITM5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFITM5
Gene Accession No. A6NNB3
Gene Alias 1110003J06Rik; AW213665; bone-restricted ifitm-like protein; bone-restricted interferon induced transmembrane protein-like protein; bone-restricted interferon-induced transmembrane protein-like protein; Bril; dispanin subfamily A member 1; DSPA1; Fragilis family member 4; fragilis4; haemopoiesis related membrane protein 1; Hrmp1; Hrtm1; Ifitm5; interferon induced transmembrane protein 5; interferon-induced transmembrane protein 5; OI5
Gene Symbols IFITM5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IFITM5. Recombinant protein control fragment (Product #RP-97567).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387733
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.