Learn More
Aat Bioquest iFluor 488 succinimidyl ester (100 ug)
SDP
Supplier:  Aat Bioquest 71023
Encompass_Preferred
Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates there are certain limitations with FITC such as severe photobleaching for microscope imaging and pH-sensitive fluorescence. This catalog is for 100 ug.
Catalog No. 50-225-0269
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.