Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFN alpha Monoclonal Antibody (7N41)
GREENER_CHOICE

Catalog No. ENM710 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENM710 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENM710 Supplier Invitrogen™ Supplier No. M710
  • $616.46
    $474.65 / Each of 1

    Save $141.81

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

M710 targets IFN alpha in ELISA applications and shows reactivity with Human samples. The M710 immunogen is recombinant human IFN alpha 2b. M710 detects IFN alpha which has a predicted molecular weight of approximately 19 kDa. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN alpha
Applications ELISA
Classification Monoclonal
Clone 7N41
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IFNA1
Gene Accession No. P01562, P01563
Gene Alias alpha-2a interferon; Gm12597; ifa; Ifa1; Ifa2; IFL; IFN; IFN alpha; IFN alpha 1/13; IFN alpha 1b; IFN α; IFN1@; IFNA; IFNA@; IFNA1; IFNA13; Ifna15; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFNA3; IFNA6; Ifnaa; IFNAalphaIFN-alpha-1/13; IFNalpha; IFN-alpha; IFN-alpha 1b; IFN-alpha1; IFN-alpha-1; IFN-alpha-1/13; IFN-alpha-2; IFN-alpha-6; IFN-alphaA; IFN-alphaD; IFN-gamma; IFNα; INFA2; interferon; interferon alpha; interferon alpha 1; Interferon alpha 13 (IFNA13); interferon alpha 15; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family gene 1; interferon alpha family, gene 2; Interferon alpha leukocyte (IFL); interferon alpha protein; interferon alpha-1; Interferon alpha1 (IFNA1); interferon alpha-1 precursor; interferon alpha-1/13; Interferon alpha-1/13 (IFN-alpha-1/13); Interferon alpha-2; Interferon alpha-A; Interferon alpha-D; Interferon alpha-D (IFN alphaD); interferon type A1/A2; interferon type A3; interferon, alpha 1; interferon, alpha 2; interferon-alpha; interferon-alpha 1; interferon-alpha1; interferon-alpha-6; leIF A; LeIF D; MGC138207; MGC138505; MGC138507; MuIFN-alpha-A; novel interferon alpha (Ifna) family member; OTTMUSG00000007655
Gene Symbols IFNA1, IFNA2
Host Species Mouse
Immunogen Recombinant human IFN alpha 2b.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3439, 3440
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.