Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFN beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PI324001 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2x10^4 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI324001 2x10^4 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI324001 Supplier Invitrogen™ Supplier No. 324001
  • $590.71
    $461.65 / Each of 1

    Save $129.06

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specificity: Neutralizes mouse interferon beta. Activity: 1.55x10^6 NU/mL. Interferon was titrated with the use of the cytopathic effect inhibition assay using L929 cells with EMCV, where 2.5 unit/mL of interferon was necessary to produce an endpoint of 50%. One neutralization unit is defined as the amount of serum required to neutralize one unit of mouse IFN beta to an endpoint of 50%. For retention of full activity, do not store above -70°C. Packaged by and distributed for PBL Assay Science.

Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN beta
Applications Neutralization
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum, PBS with no preservative
Gene Ifnb1
Gene Accession No. P01575
Gene Alias Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1
Gene Symbols Ifnb1
Host Species Rabbit
Immunogen Recombinant mouse interferon beta.
Quantity 2x10^4 units
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15977
Target Species Mouse
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
The antibody I purchased from you says ''concentration not determined'' on the product label and on the product page. How do I determine the concentration?

Some antibodies are provided in non-purified formats, such as ascites fluid, tissue culture supernatant, or whole serum. These formats contain multiple proteins, which can interfere with accurate protein quantification. Traditional methods like A280 measurement or protein assays such as BCA quantify total protein but do not provide a specific concentration for the antibody in question.

Literature, such as "Antibodies" by Harlow & Lane (1988) (https://antibodiesmanual.org/index.php) (Table 8.2 - Storing and Purifying Antibodies), suggests approximate concentrations of antibodies in these solutions, as shown below. However, please note that these are rough estimates, and actual concentrations can vary based on several factors.

Serum
Total antibody concentration: 10 mg/mL
Specific antibody concentration: 0.1 to 1 mg/mL (a maximum of 10% of the antibody population is expected to be the specific antibody of interest)

Tissue Culture Supernatant (10% FBS)
Total antibody concentration: 1 mg/mL
Specific antibody concentration: 0.05 mg/mL

Tissue Culture Supernatant (serum-free)
Total antibody concentration: 0.01 to 0.05 mg/mL
Specific antibody concentration: 0.01 to 0.05 mg/mL

Ascites
Total antibody concentration: 1 to 10 mg/mL
Specific antibody concentration: 0.9 to 9 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.