Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IFN-gamma Antibody Pair
SDP

Catalog No. NB468711 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB468711 15 Plates
NB468710 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB468711 Supplier Novus Biologicals™ Supplier No. NBP33075715PLATES
Only null left
Add to Cart
Add to Cart

Antibody Pair

Specifications

Assay Range 31.25-2000 pg/mL (example only; lot dependent)
Assay Sensitivity 31.25 pg/mL (example only; lot dependent)
Conjugate Unconjugated
Product Type Sandwich ELISA
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse IFN-gamma.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3458
Gene Symbol IFNG
Quantity 15 Plates
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.