Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IFN-gamma ELISA Development Kit
SDP

Catalog No. NB397893 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit (for 6 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397893 1 Kit (for 6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397893 Supplier Novus Biologicals™ Supplier No. NBP311746

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

ELISA Development Kit

Specifications

Assay Range 12 - 1200 pg/mL
Product Type ELISA Development Kit
Sample Type Serum, Plasma, Cell Culture Supernatant
For Use With (Application) ELISA
Gene ID (Entrez) 3458
Gene Symbol IFNG
Quantity 1 Kit (for 6 Plates)
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Target Ferret
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.