Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0291510
Encompass_Preferred
Human Interferon gamma (hIFN-gamma) is amacrophage‐activating factor and the lone member of Interferon type II.The active form of IFN-gamma is an antiparallel dimer that interacts with the receptor IFN-gammaR1 and sets off IFN-gamma/JAK/STAT pathway. IFN-gamma signaling does diverse biological functions primarily related to host defense and immune regulation including antiviral and antibacterial defense apoptosis inflammation and innate and acquired immunity. While IFN-gamma–induced inflammatory cascade summons a variety of immune‐related cell types such as macrophages natural killer (NK) cells and cytotoxic T lymphocytes (CTLs) IFN-gamma is also implicated in resistance to NK cell and CTL responses and in immune escape in a variety of cancers.
Catalog No. 50-252-9958
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.