Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFN gamma Monoclonal Antibody (AN-18), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5013680
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-136-80 50 μg
50-136-81 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-136-80 Supplier Invitrogen™ Supplier No. 14731381
  • $119.00
    $89.65 / Each of 1

    Save $29.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The AN-18 antibody reacts with mouse interferon-gamma (IFN gamma), a 15-20 kDa cytokine secreted by Th1 cells. Applications Reported: The AN-18 antibody has been reported for use in immunoprecipitation, ELISPOT, and ELISA. (Please use Functional Grade purified AN-18, cat. 16-7313, for ELISPOT Capture.). Applications Tested: The AN-18 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse IFNg (Interferon-gamma, IFN-g) in combination with the biotin R4-6A2 (13-7312) antibody for detection and recombinant mouse IFNg (39-8311) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 0.25-1 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 200 pg/mL - 1.6 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN gamma
Applications Immunoprecipitation, Immunostaining
Classification Monoclonal
Clone AN-18
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene IFNG
Gene Accession No. P01580
Gene Alias BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-g
Gene Symbols IFNG
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15978
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.