Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFN gamma Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P11950UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P11950UG 50 μg
500P1191MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 500P11950UG Supplier Invitrogen™ Supplier No. 500P11950UG
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MHGTVIESLE SLNNYFNSSG IDVEEKSLFL DIWRNWQKDG DMKILQSQII SFYLRLFEVL KDNQAISNNI SVIESHLITT FFSNSKAKKD AFMSIAKFEV NNPQVQRQAF NELIRVVHQL LPESSLRKRK RSRC. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IFN-gamma. Anti-Murine IFN-gamma-specific antibody was purified by affinity chromatography employing an immobilized Murine IFN-gamma matrix. Sandwich ELISA: To detect Murine IFN-gamma by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine IFN-gamma (500-P119Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine IFN-gamma. Western Blot: To detect Murine IFN-gamma by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Murine IFN-gamma is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN gamma
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IFNG
Gene Accession No. P01580
Gene Alias BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-g
Gene Symbols IFNG
Host Species Rabbit
Immunogen E.coli-derived Recombinant Murine IFN-gamma.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15978
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.