Learn More
Genscript Corporation Antibody Protein Proteins Proteins 100 UG 1/EA
Supplier:  Genscript Corporation Z03584100
Encompass_Preferred
Interferon-gamma (IFN-gamma) also known as Type II interferon or immune interferon is a cytokine produced primarily by T-lymphocytes and natural killer cells. The protein shares no significant homology with IFN-beta or the various IFN-Alpha family proteins. Mature IFN-gamma exists as noncovalently-linked homodimers. It shares high sequence indentity with mouse IFN-gamma (86 %). IFN-gamma was originally characterized based on its antiviral activities. The protein also exerts antiproliferative immunoregulatory and proinflammatory activities and is thus important in host defense mechanisms. IFN-gamma induces the production of cytokines upregulates the expression of class I and II MHC antigens Fc receptor and leukocyte adhesion molecules. It modulates macrophage effector functions influences isotype switching and potentiates the secretion of immunoglobulins by B cells. Additionally IFN-gamma augments TH1 cell expansion and may be required for TH1 cell differentiation.
Catalog No. 50-252-9604
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.