Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNA14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103422
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5103422 100 μL
1 options

Catalog No. PIPA5103422

Supplier: Invitrogen™ PA5103422

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFN14.

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNA14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFNA14
Gene Accession No. P01570
Gene Alias IFNA14; IFN-alpha-14; IFN-alphaH; Interferon; interferon alpha 14; interferon alpha-14; Interferon alpha-H; interferon lambda-2-H; interferon, alpha 14; leIF H; LEIF2H; OTTMUSG00000007662
Gene Symbols IFNA14
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFNA14(Accession P01570), corresponding to amino acid residues L105-M155.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3448
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.