Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNAR1 Monoclonal Antibody (MAR1-5A3), PE, eBioscience™
GREENER_CHOICE

Catalog No. 501122359
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-2359 Supplier Invitrogen™ Supplier No. 12594582
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The MAR1-5A3 monoclonal antibody reacts with mouse IFN alpha/beta receptor subunit 1 (IFNAR-1). The type I IFN receptor subunit 1 is co-expressed with IFNAR-2 on nearly all cells and make up the heterodimeric receptor complex that binds to all type I IFNs, IFN alpha and IFN beta. Type I IFNs are a group of structurally and functionally related proteins that demonstrate antiviral, antiparasitic, and antiproliferative activities. Applications Reported: This MAR1-5A3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MAR1-5A3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system (Biron,2001, Kirkwood, 2002). These cytokines produce their effects by binding to the Type 1 Interferon-& Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1 (Kumar et al., 2003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNAR1
Applications Flow Cytometry
Classification Monoclonal
Clone MAR1-5A3
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Ifnar1
Gene Accession No. P33896
Gene Alias alpha-type antiviral protein; AVP; beta-type antiviral protein; CD118; CRF2-1; cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; H3K1; Ifar; IFN R1; IFNalpha/beta R1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFNalpha/betaR1; IFN-alpha/betaR1; IFN-alpha/betaRalpha; IFNalpha/beta-Ralpha; IFN-alpha-REC; IFNAR; Ifnar1; IFNBR; IFNR1; IFN-R-1; Ifrc; INF-a receptor; Infar; interferon (alpha and beta) receptor 1; interferon (alpha, beta and omega) receptor 1; interferon alpha and beta receptor subunit 1; interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha/beta receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; type I interferon receptor 1
Gene Symbols Ifnar1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15975
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.