Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNAR2 Mouse anti-Human, Clone: 7, Invitrogen™

Catalog No. PIMA529341
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529341 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529341 Supplier Thermo Scientific Supplier No. MA529341

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNAR2
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 7
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IFNAR2
Gene Accession No. P48551
Gene Alias interferon (alpha, beta and omega) receptor 2
Gene Symbols IFNAR2
Host Species Mouse
Immunogen Recombit Human IFNAR2 protein
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3455
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.