Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNGR1 Mouse anti-Human, Clone: 1H3A4, Proteintech
SDP

Catalog No. p-7213168 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7119 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7119 Supplier Proteintech Group Inc Supplier No. 667041IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.

Specifications

Antigen IFNGR1
Applications Western Blot
Classification Monoclonal
Clone 1H3A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene IFNGR1
Gene Accession No. P15260
Gene Alias CD119, CDw119, FLJ45734, IFN gamma R1, IFN gamma receptor 1, IFNGR, IFNGR1, interferon gamma receptor 1
Gene Symbols IFNGR1
Host Species Mouse
Immunogen IFNGR1 Fusion Protein Ag27085
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3459
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.