Learn More
Hemosure™ iFOB Test Kits

Description
- CLIA Complexity: Waived
- CPT Codes: G0328QW (colorectal cancer screening); 82274QW (detection of blood in feces form)
- Sensitivity: 50ng hHB/mL
- Specificity: over 96%
- Useful aid in the diagnosis and therapy of gastrointestinal disorders
- Patient-friendly for use in physicians' offices, labs, hospitals, clinics, or homes
- As test sample flows up through absorbent device, the labeled antibody-dye conjugate binds to the hemoglobin in specimen, forming an antibody-antigen complex
- Results within five minutes
- Positive results: one band in the control (C) region, one band in the test (T) region
- Negative results: only one band in the control (C) region
- Low false positive results
- Reaction components: citric acid, monoclonal and polyclonal antibody, mouse anti-HB antibodies 1 and 2, and goat anti-mouse IgG antibody
If home kit mailers are not needed, they can be excluded from the order. Please contact your Fisher HealthCare account manager for details.
Order Info
The 10-test kit contains 10 collection bottles and 10 home kit mailers; 30-test kit contains 50 collection bottles and 50 home kit mailers.
Specifications
Specifications
| Content And Storage | 30 Fecal occult Blood test cassettes, 30 collection tubes |
| CPT Code | G0328QW, 82274QW |
| Clia Complexity | Waived |
| Type | iFOB Test Kit |
| For Use With (Application) | Diagnosis and therapy of gastrointestinal disorders |
| Includes | 30 tests, 30 tubes, no mailers |
| Quantity | 30 Pack |
| Test Time | 5 min. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.