Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFRD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103431
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5103431 100 μL
1 options

Catalog No. PIPA5103431

Supplier: Invitrogen™ PA5103431

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFRD2.

IFRD2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFRD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFRD2
Gene Accession No. Q12894
Gene Alias 1810034A24Rik; AA470234; AI838527; IFNRP; IFRD2; interferon related developmental regulator 2; interferon-related developmental regulator 2; interferon-related protein; Protein SKMC15; SKMc15; SM15
Gene Symbols IFRD2
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFRD2(Accession Q12894), corresponding to amino acid residues L358-E408.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15983, 7866
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.