Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT88 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544087 Supplier Invitrogen™ Supplier No. PA544087
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EIREQRIKSG RDGSGGSRGK REGSASGDSG QNYSASSKGE RLSARLRALP Sequence homology: Dog: 92%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 85%; Rabbit: 86%; Rat: 86%.

This gene encodes a member of the tetratrico peptide repeat family. Mutations of a similar gene in mouse can cause polycystic kidney disease. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT88
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IFT88
Gene Accession No. Q13099
Gene Alias AW552028; D13S1056E; DAF19; fb37b11; flexo; fxo; HGNC:20606; hTg737; Ift88; intraflagellar transport 88; intraflagellar transport 88 homolog; intraflagellar transport protein; intraflagellar transport protein 88; intraflagellar transport protein 88 homolog; MGC26259; orpk; osm-5; oval; ovl; polaris; polaris homolog; polaris; axonemal transport protein; probe hTg737 (polycystic kidney disease, autosomal recessive); recessive polycystic kidney disease protein Tg737; Recessive polycystic kidney disease protein Tg737 homolog; RP11-172H24.2; testicular tissue protein Li 93; tetratricopeptide repeat domain 10; tetratricopeptide repeat protein 10; TG737; Tg737 protein; Tg737Rpw; TgN(Imorpk)737Rpw; TgN737Rpw; TPR repeat protein 10; transgene insert site 737, insertional mutation, polycystic kidney disease; Ttc10; wu:fb37b11; xx:zah8127
Gene Symbols IFT88
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human IFT88.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8100
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.