Learn More
Novus Biologicals™ IFTAP Overexpression Lysate

Description
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC28417 and based on accession number NM_138787. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens chromosome 11 open reading frame 74 (C11orf74), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Target | IFTAP |
| Synonym | chromosome 11 open reading frame 74, FLJ38678, HEPIS, hypothetical protein LOC119710, NWC, Protein HEPIS |
| Protein State | Native |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.