Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig Goat anti-Rabbit, Human ads, R-PE, Polyclonal, Southern Biotech™

Goat Polyclonal Secondary Antibody
Supplier: Southern Biotechnology Associates 401009S
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
| Ig | |
| Polyclonal | |
| R-PE | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Rabbit | |
| IgG |
| Flow Cytometry, ELISA | |
| 0.5mg/mL | |
| Supplied in PBS/NaN3 and a stabilizing agent | |
| Rabbit IgG | |
| 0.25 mg | |
| Secondary | |
| Store at 2-8°C; Protect from light; Do not freeze |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction