Learn More
Invitrogen™ Rat Ig Isotyping Instant ELISA™ Kit
Description
The Rat Immunoglobulin Isotyping ELISA qualitatively detects Rat Ig isotypes in rat hybridoma cell culture supernatants.
Principle of the method Biotinylated monoclonal anti-rat isotype specific or light chain specific capture antibodies bind to the Streptavidin coated plate. Ig Isotype present in the positive control or sample binds to the specific capture antibodies and is simultaneously bound by the HRP-conjugated detector antibody. Following incubation unbound reagents and sample components are removed during a wash step, and substrate solution reactive with HRP is added to the wells. A colored product is formed in proportion to the amount of soluble rat Ig (H+L) present in the sample.
Rigorous validation Each manufactured lot of this ELISA kit is quality tested. See manual for more information on validation.
Specifications
Specifications
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Supernatant |
| Target Species | Rat |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Kit Contents | Pre-coated 96 well Microplate coated with Streptavidin, Biotinylated monoclonal anti-rat isotype or light chain-specific capture antibodies (Isotype specific: anti-IgG1, IgG2a, IgG2b, IgG2c, IgA, IgM; Light chain-specific: kappa or lambda), HRP-Conjugated Anti-rat lg (H&L) monoclonal antibody, Sample Diluent, lyophilized, Rat Positive Control: Rat IgG1, IgG2a, IgG2b, IgG2c, IgA, IgM, kappa and lambda isotype control mix (lyophilized), Wash Buffer Concentrate 20x (PBS with 1% Tween-20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Plate Covers |
| Label or Dye | HRP |
| Quantity | 2 x 96 Tests |
| Regulatory Status | RUO |
| Sample Volume | Supernatant 10 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.