Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IgA Monoclonal Antibody (11-44-2), Biotin, eBioscience™
GREENER_CHOICE

Catalog No. 5012115
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-121-15 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-121-15 Supplier Invitrogen™ Supplier No. 13599482
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The 11-44-2 monoclonal antibody reacts with the alpha heavy chain of mouse IgA. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgM. IgA is expressed as a membrane-bound form as well as a soluble dimer. Fluorochrome conjugated 11-44-2 is used as a detection secondary for mouse IgA. Applications Reported: The 11-44-2 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 11-44-2 antibody has been tested by flow cytometric analysis to detect a mouse IgA primary antibody. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

Mouse IgA is an immunoglobulin A antibody that is produced by B cells in response to an antigen. It is a crucial component of the mucosal immune system, which protects the body from pathogens that enter through mucosal surfaces such as the respiratory, gastrointestinal, and urogenital tracts. Mouse IgA is composed of two heavy chains and two light chains, and it is the most abundant immunoglobulin in mucosal secretions. It plays a vital role in neutralizing pathogens and preventing their attachment to mucosal surfaces. Mouse IgA is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgA
Applications Flow Cytometry
Classification Monoclonal
Clone 11-44-2
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Igha
Gene Accession No. P01878
Gene Alias IgA; Igh-2; Igha; immunoglobulin heavy constant alpha; ImmunoglobulinA
Gene Symbols Igha
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238447
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.