Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat IgA Uncoated ELISA Kit

Catalog No. 885048088
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-504-8088 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-504-8088 Supplier Invitrogen™ Supplier No. 885048088

ELISA

The Rat Immunoglobulin A (IgA) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat IgA. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Rat IgA is an essential immunoglobulin isotype that is predominantly present in mucosal secretions like colostrum, saliva, and tears. It acts as a crucial defense mechanism against pathogens that enter the body through mucosal surfaces. Rat IgA is made up of two heavy chains and two light chains, with a molecular weight of approximately 170 kDa. It is produced by B cells in response to antigenic stimulation and has the ability to bind to various antigens, including bacteria, viruses, and toxins. Rat IgA functions by neutralizing these pathogens and preventing their attachment to mucosal surfaces, which helps to reduce the risk of infection. It also activates complement and recruits immune cells to the site of infection, further enhancing the immune response. Rat IgA is widely used as a research tool in immunology and infectious disease studies and has been produced for therapeutic applications such as in the treatment of inflammatory bowel disease and mucosal infections. Overall, Rat IgA is a critical component of the immune system and plays a crucial role in protecting the body against mucosal infections.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 16 to 1,000 ng/mL
Assay Sensitivity 16 ng/ml
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Rat
For Use With (Equipment) Colorimetric Microplate Reader
Kit Contents Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Rat IgA protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 10 μL, Serum 10 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Rat IgA
Test Time 1 hrs 45 mins
Total Assay Time 24 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.