Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD Antibody (RM123), Biotin, R&D Systems™
SDP

Catalog No. NBP262026B Shop All Novus Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262026B 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262026B Supplier R&D Systems Supplier No. NBP262026B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

IgD Monoclonal specifically detects IgD in Human samples. It is validated for ELISA, Immunohistochemistry, Sandwich ELISA Capture, Sandwich ELISA Detection.

Specifications

Antigen IgD
Applications ELISA, Immunohistochemistry, Sandwich ELISA Capture, Sandwich ELISA Detection
Classification Monoclonal
Clone RM123
Concentration 1 mg/mL
Conjugate Biotin
Dilution Flow Cytometry, ELISA, Immunohistochemistry 0.5 - 2 ug/mL, Immunocytochemistry/Immunofluorescence, Immunocytochemistry 0.5 - 2 ug/mL
Formulation PBS, 50% Glycerol, 1% BSA with 0.09% Sodium Azide
Gene Alias IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Gene Symbols IGHD
Host Species Rabbit
Immunogen Human IgD
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3495
Test Specificity This antibody reacts to human IgD. No cross reactivity with human IgG, IgM, IgA, or IgE.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.