Learn More
Invitrogen™ Human IgD Monoclonal Antibody (IA6-2), NovaFluor™ Violet 745, eBioscience™
Description
This IA6-2 monoclonal antibody reacts with the heavy chain of human immunoglobulin D (IgD). This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding. This product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer. Applications Tested This IA6-2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 0.4 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Master Mixes Master mixes of NFs should be made at 2-8 °C and may be made up to 4 hours ahead of time. We do not recommend storing master mixes containing NovaFluor conjugates overnight or longer. Whole Blood Compatibility When utilizing whole blood (as opposed to density-gradient-purified PBMC), we recommend lysing red blood cells in bulk prior to staining with NovaFluor conjugates. See the Bulk Lysis of Human Whole Blood protocol here. Staining of whole blood with NovaFluor conjugates followed by lysis of red blo...
Specifications
Specifications
| Antigen | Human IgD |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | IA6-2 |
| Concentration | 0.4 μg/Test |
| Conjugate | NovaFluor Violet 745 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | IGHD |
| Gene Accession No. | P01880 |
| Gene Alias | Ig delta chain constant region; IGHD; IGHD1; IGHDELTA; immunoglobulin heavy constant delta |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.