Learn More
Enzo Life Sciences IgE (non-immune) (human), (biotin conjugate) (150 µg)
SDP
Supplier:  Enzo Life Sciences BPDDIAHE1B015
Encompass_Preferred
Synonyms: Immunoglobulin E. Source: Purified human IgE with kappa light chains produced in vitro from a monoclonal hybridoma. Applications: ICC, FUNC. Purification: Protein L purified. The original antibody-producing B cells were obtained from a healthy donor tested negative by US-FDA approved tests against HIV, HCV and Hepatitis B. Although the antibody comprising BPD-DIA-HE1B is purified from a monoclonal hybridoma line derived from the antibody-producing donor cell, it must be handled as potentially infectious as all human material. As the IgE comes from a monoclonal cell line, there is no contamination of antibodies of other isotypes. The remaining contaminants are mainly components from fetal bovine serum. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.14M sodium chloride and 15mM sodium azide.
Catalog No. 50-210-0405
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.