Learn More
Description
MA1-10854 detects IGF-1 Receptor from human samples. MA1-10854 has been successfully used in Western blot applications. Dilute in PBS or medium which is identical to that used in the assay system. The MA1-10854 immunogen is purified human placental IGF-1 receptors.
Specifications
Specifications
| Antigen | IGF1R (CD221) |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1H7 |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | IGF1R |
| Gene Accession No. | P08069 |
| Gene Alias | A330103N21Rik; CD221; CD221 antigen; D930020L01; hyft; IGF 1 receptor; IGF 1R; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; Insulin like growth factor 1 receptor precursor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
