Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

$250.00 - $2175.00

Specifications

Antigen IGFBP-1
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
500P228BT25
Encompass_Preferred
View Documents
Invitrogen™
500P228BT25UG
25 μg
Each for $250.00
Only null left
Add to Cart
 
50-271-8593
Encompass_Preferred
View Documents
Invitrogen™
500P228BT500UG
500 μg
Each for $2,175.00
Only null left
Add to Cart
 
Description

Description

AA Sequence of recombinant protein: MAPWQCAPCS AEKLALCPPV SASCSEVTRS AGCGCCPMCA LPLGAACGVA TARCARGLSC RALPGEQQPL HALTRGQGAC VQESDASAPH AAEAGSPESP ESTEITEEEL LDNFHLMAPS EEDHSILWDA ISTYDGSKAL HVTNIKKWKE PCRIELYRVV ESLAKAQETS GEEISKFYLP NCNKNGFYHS RQCETSMDGE AGLCWCVYPW NGKRIPGSPE IRGDPNCQIY FNVQN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IGF-BP1. Anti-Human IGF-BP1-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hIGF-BP1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IGF-BP1 (500-P228) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hIGF-BP1. Western Blot: To detect hIGF-BP1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIGF-BP1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P228BT-1MG will be provided as 2 x 500 μg

This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
Specifications

Specifications

IGFBP-1
ELISA, Western Blot
Biotin
Rabbit
Human
P08833
3484
E.coli-derived Recombinant Human IGF-BP1.
Antigen affinity chromatography
IGFBP1
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
AFBP; alpha-pregnancy-associated endometrial globulin; amniotic fluid binding protein; Binding protein 25; Binding protein 26; Binding protein 28; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; hIGFBP 1; hIGFBP1; hIGFBP-1; IBP 1; IBP1; IBP-1; IGF BP25; IGFBA; IGF-binding protein 1; IGFBP; IGFBP 1; Igfbp1; Igfbp-1; IGF-BP25; Insulin like growth factor binding protein; insulin like growth factor binding protein 1; insulin-like growth factor binding protein 1; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 1 PRECURSOR (IGFBP-1) (IBP-1) (IGF-BINDING PROTEIN 1); insulin-like growth factor-binding protein 1; Placental protein 12; PP 12; PP12
IGFBP1
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.