Learn More
BioVendor, LLC Igfbp-4 Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC REH005R
Encompass
The BioVendor Human IGFBP-4 ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IGFBP-4 in serum, plasma, and cell culture supernatants. This assay employs an antibody specific for human IGFBP-4 coated on a 96well plate. Standards and samples are pipetted into the wells and IGFBP-4 present in a sample is bound to the wells by the immobilized antibody. The wells are washed and biotinylated anti-human IGFBP-4 antibody is added. After washing away unbound biotinylated antibody, HRP-conjugated streptavidin is pipetted to the wells. The wells are again washed, a TMB substrate solution is added to the wells and color develops in proportion to the amount of IGFBP-4 bound. The Stop Solution changes the color from blue to yellow, and the intensity of the color is measured at 450 nm. One kit (96 wells). Calibration range: 0.41-100ng/ml. Limit of detection: 250pg/ml. Other names: Insulin-like growth factor binding protein-4.
Catalog No. 50-240-6380
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.