Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGFBP2 Recombinant Rabbit Monoclonal Antibody (106)
GREENER_CHOICE

Catalog No. PIMA531156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531156 Supplier Invitrogen™ Supplier No. MA531156
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse IGFBP-2/IGFBP2.

IGFBP2 (insulin-like growth factor binding protein 2) is a member of the ISGBP family which bind various IGFs. IGFBP2 is overexpressed in a wide spectrum of cancers, including glioma, prostate cancer, synovial sarcoma, neuroblastoma, colon cancer, adrenocortical cancer, lung cancer, Wilms' tumor, and hepatoblastoma. The overexpression of IGFBP2 also correlates with the aggressiveness of some tumors. IGFBP2 activates the expression of matrix metalloprotease 2, which contributes to cell invasiveness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP2
Applications ELISA
Classification Recombinant Monoclonal
Clone 106
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IGFBP2
Gene Accession No. P47877
Gene Alias AI255832; BP2; BRL-BP; IBP2; IBP-2; IGF binding protein 2; IGF-binding protein 2; IGFBP; IGFBP2; IGFBP-2; IGF-BP53; ILGFBPA; Insulin like growth factor binding protein; insulin like growth factor binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36kDa; insulin-like growth factor-binding protein 2; mIGFBP-2; OTTHUMP00000207753
Gene Symbols IGFBP2
Host Species Rabbit
Immunogen Recombinant Mouse IGFBP-2/IGFBP2 Protein (Met1-Gln305).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16008
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.