Learn More
AdipoGen IGFL3 human rec
SDP
Supplier:  AdipoGen AG40B00903010
Encompass_Preferred
Recombinant Protein. Human IGFL3 aa 25-125 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Lyophilized. Contains PBS. Insulin-growth factor-like gene family is a new family of proteins consisting of four proteins in humans IGFL1 to 4 and one in mice mIGFL. mIGFL is expressed in normal skin in mice and further upregulated during inflammation responses in skin or after skin wounding. In human only IGFL1 expression is increased in psoriatic skin samples. mIGFL and human IGFL1 and 3 interact with specificity and high affinity to a novel receptor named IGF-like family receptor 1 formerly TMEM-149. Analysis of the amino acid sequence of IGFLR1 indicated that this receptor is likely a novel member of the TNF-R family. IGFLR1 is expressed most abundantly on mouse T cells, suggesting that mIGFL and IGFL1 produced in the skin may potentially exert regulatory functions on T cell responses.
Catalog No. 50-256-2924
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.