Learn More
Diagnostic Biosystems IGG IgG
Supplier:  Diagnostic Biosystems RP023
Encompass_Preferred
This antibody reacts with γ-chains of human IgG. Nonspecific antibodies have been removed by solid-phase adsorption.
Catalog No. 50-238-5534
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.