Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ IgG Affinity Gel

Catalog No. ICN55961 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55961 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55961 Supplier MP Biomedicals™ Supplier No. 0855961
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IgG Affinity Gel is prepared by reaction of purified IgG to activated Sepharose™ 4B gel.

  • Product is rabbit IgG (whole molecule) covalently coupled to Sepharose-4B
  • Packed in a polystyrene chromatography column
TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 2.3 mg Antibody/mL
Quantity 2 mL
Content And Storage 2°C to 8°C
For Use With (Application) For affinity purification of antibodies to Rabbit IgG or to remove these antibodies from an antiserum preparation

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.