Learn More
Alpco Ltd Canine IgG ELISA
SDP
Supplier:  Alpco Ltd 41IGGCAE01
Encompass
The Canine IgG ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgG in biological samples of dogs. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3592
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.